Order Entry
Glass crimp top headspace vials, 10 ml, level 2 high-throughput applications, SureSTART™
Glass crimp top headspace vials, 10 ml, level 2 high-throughput applications, SureSTART™
Certificates
undefined
Glass crimp top headspace vials, 10 ml, level 2 high-throughput applications, SureSTART™
Vials
Product Information
  • Catalog #
    Supplier #
    Capacity
    Base
    Ø×H
    Material
    Colour
    Availability
    Price
  • Crimp vial, headspace
    Crimp vial, headspace
    548-2703
    6ACV10-1R
    10,00 ml
    Beveled edge round bottom
    22,5×46 mm
    Glass
    Clear
    Log in for availability
    Log in for availability
    1 * 100 KS
    1 * 100 KS - 1 730,00 Kč
    1 730,00 Kč
    1 * 100 KS
       1 730,00 Kč
    Supplier Article Number: 6ACV10-1R
    Local Article Number: HYPE6ACV10-1R
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Crimp vial, headspace
    Crimp vial, headspace
    548-2706
    6ACV10-2R
    10,00 ml
    Beveled edge round bottom
    22,5×46 mm
    Glass
    Amber
    Log in for availability
    Log in for availability
    1 * 100 KS
    1 * 100 KS - 2 320,00 Kč
    2 320,00 Kč
    1 * 100 KS
       2 320,00 Kč
    Supplier Article Number: 6ACV10-2R
    Local Article Number: HYPE6ACV10-2R
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Crimp vial, headspace
    Crimp vial, headspace
    548-2705
    6ACV10-1RT
    10,00 ml
    Beveled edge round bottom
    22,5×46 mm
    Glass
    Clear
    Log in for availability
    Log in for availability
    1 * 1.000 KS
    1 * 1.000 KS - 16 420,00 Kč
    16 420,00 Kč
    1 * 1.000 KS
       16 420,00 Kč
    Supplier Article Number: 6ACV10-1RT
    Local Article Number: HYPE6ACV10-1RT
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Crimp vial, headspace
    Crimp vial, headspace
    548-2708
    6ACV10-2RT
    10,00 ml
    Beveled edge round bottom
    22,5×46 mm
    Glass
    Amber
    Log in for availability
    Log in for availability
    1 * 1.000 KS
    1 * 1.000 KS - 22 020,00 Kč
    22 020,00 Kč
    1 * 1.000 KS
       22 020,00 Kč
    Supplier Article Number: 6ACV10-2RT
    Local Article Number: HYPE6ACV10-2RT
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Crimp vial, headspace
    Crimp vial, headspace
    548-2704
    6ACV10-1RF
    10,00 ml
    Beveled edge round bottom
    22,5×46 mm
    Glass
    Clear
    Log in for availability
    Log in for availability
    1 * 5.000 KS
    1 * 5.000 KS - 77 980,00 Kč
    77 980,00 Kč
    1 * 5.000 KS
       77 980,00 Kč
    Supplier Article Number: 6ACV10-1RF
    Local Article Number: HYPE6ACV10-1RF
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Crimp vial, headspace
    Crimp vial, headspace
    548-2707
    6ACV10-2RF
    10,00 ml
    Beveled edge round bottom
    22,5×46 mm
    Glass
    Amber
    Log in for availability
    Log in for availability
    1 * 5.000 KS
    1 * 5.000 KS - 104 570,00 Kč
    104 570,00 Kč
    1 * 5.000 KS
       104 570,00 Kč
    Supplier Article Number: 6ACV10-2RF
    Local Article Number: HYPE6ACV10-2RF
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
Caps
Product Information
  • Catalog #
    Seq
    Description
    Hardness
    Septum
    Thickness
    Cap colour
    Cap Size
    Material
    Availability
    Price
  • 20
    Crimp cap
    45 shore
    White silicone/beige PTFE
    3,2 mm
    Silver
    20 mm
    Aluminium
    Log in for availability
    Log in for availability
    1 * 100 KS
    1 * 100 KS - 2 280,00 Kč
    2 280,00 Kč
    Supplier Article Number: 6ACC20-ST1
    Local Article Number: HYPE6ACC20-ST1
Product Information
  • Catalog #
    Seq
    Description
    Hardness
    Septum
    Thickness
    Cap colour
    Cap Size
    Material
    Availability
    Price
  • 13
    Crimp cap
    50 shore
    Black butyl/grey PTFE, molded pharmafix
    3,0 mm
    Silver
    20 mm
    Aluminium
    Log in for availability
    Log in for availability
    1 * 100 KS
    1 * 100 KS - 3 540,00 Kč
    3 540,00 Kč
    Supplier Article Number: 6ACC20-PFT
    Local Article Number: HYPE6ACC20-PFT
Product Information
  • Catalog #
    Seq
    Description
    Hardness
    Septum
    Thickness
    Cap colour
    Cap Size
    Material
    Availability
    Price
  • 22
    Crimp cap
    45 shore
    White silicone/beige PTFE
    3,2 mm
    Silver
    20 mm
    Aluminium
    Log in for availability
    Log in for availability
    1 * 1.000 KS
    1 * 1.000 KS - 21 880,00 Kč
    21 880,00 Kč
    Supplier Article Number: 6ACC20-ST1T
    Local Article Number: HYPE6ACC20-ST1T
Product Information
  • Catalog #
    Seq
    Description
    Hardness
    Septum
    Thickness
    Cap colour
    Cap Size
    Material
    Availability
    Price
  • 11
    Crimp cap
    50 shore
    Black butyl/grey PTFE, molded pharmafix
    3,0 mm
    Silver
    20 mm
    Aluminium
    Log in for availability
    Log in for availability
    1 * 1.000 KS
    1 * 1.000 KS - 33 580,00 Kč
    33 580,00 Kč
    Supplier Article Number: 6ACC20-PFTT
    Local Article Number: HYPE6ACC20-PFTT
Product Information
  • Catalog #
    Seq
    Description
    Hardness
    Septum
    Thickness
    Cap colour
    Cap Size
    Material
    Availability
    Price
  • 21
    Crimp cap
    45 shore
    White silicone/beige PTFE
    3,2 mm
    Silver
    20 mm
    Aluminium
    Log in for availability
    Log in for availability
    1 * 5.000 KS
    1 * 5.000 KS - 102 800,00 Kč
    102 800,00 Kč
    Supplier Article Number: 6ACC20-ST1F
    Local Article Number: HYPE6ACC20-ST1F
Product Information
  • Catalog #
    Seq
    Description
    Hardness
    Septum
    Thickness
    Cap colour
    Cap Size
    Material
    Availability
    Price
  • 11
    Crimp cap
    50 shore
    Black butyl/grey PTFE, molded pharmafix
    3,0 mm
    Silver
    20 mm
    Aluminium
    Log in for availability
    Log in for availability
    1 * 5.000 KS
    1 * 5.000 KS - 148 270,00 Kč
    148 270,00 Kč
    Supplier Article Number: 6ACC20-PFTF
    Local Article Number: HYPE6ACC20-PFTF
Product Information
  • Catalog #
    Seq
    Description
    Hardness
    Septum
    Thickness
    Cap colour
    Cap Size
    Material
    Availability
    Price
  • 17
    Crimp cap, pressure release
    45 shore
    White silicone/beige PTFE
    3,0 mm
    Silver
    20 mm
    Aluminium
    Log in for availability
    Log in for availability
    1 * 100 KS
    1 * 100 KS - 1 910,00 Kč
    1 910,00 Kč
    Supplier Article Number: 6APRCC20-ST1
    Local Article Number: HYPE6APRCC20-ST1
Product Information
  • Catalog #
    Seq
    Description
    Hardness
    Septum
    Thickness
    Cap colour
    Cap Size
    Material
    Availability
    Price
  • 8
    Magnetic crimp cap
    45 shore
    Blue silicone/clear PTFE, soft
    3,0 mm
    Gold
    20 mm
    Steel
    Log in for availability
    Log in for availability
    1 * 100 KS
    1 * 100 KS - 4 330,00 Kč
    4 330,00 Kč
    Supplier Article Number: 6AMCC20-ST3
    Local Article Number: HYPE6AMCC20-ST3
Product Information
  • Catalog #
    Seq
    Description
    Hardness
    Septum
    Thickness
    Cap colour
    Cap Size
    Material
    Availability
    Price
  • 7
    Crimp cap, pressure release
    45 shore
    White silicone/beige PTFE
    3,0 mm
    Silver
    20 mm
    Aluminium
    Log in for availability
    Log in for availability
    1 * 1.000 KS
    1 * 1.000 KS - 17 910,00 Kč
    17 910,00 Kč
    Supplier Article Number: 6APRCC20-ST1T
    Local Article Number: HYPE6APRCC20-ST1T
Product Information
  • Catalog #
    Seq
    Description
    Hardness
    Septum
    Thickness
    Cap colour
    Cap Size
    Material
    Availability
    Price
  • 16
    Magnetic crimp cap
    45 shore
    Blue silicone/clear PTFE, soft
    3,0 mm
    Gold
    20 mm
    Steel
    Log in for availability
    Log in for availability
    1 * 1.000 KS
    1 * 1.000 KS - 41 050,00 Kč
    41 050,00 Kč
    Supplier Article Number: 6AMCC20-ST3T
    Local Article Number: HYPE6AMCC20-ST3T
Product Information
  • Catalog #
    Seq
    Description
    Hardness
    Septum
    Thickness
    Cap colour
    Cap Size
    Material
    Availability
    Price
  • 14
    Crimp cap, pressure release
    45 shore
    White silicone/beige PTFE
    3,0 mm
    Silver
    20 mm
    Aluminium
    Log in for availability
    Log in for availability
    1 * 5.000 KS
    1 * 5.000 KS - 85 070,00 Kč
    85 070,00 Kč
    Supplier Article Number: 6APRCC20-ST1F
    Local Article Number: HYPE6APRCC20-ST1F
Product Information
  • Catalog #
    Seq
    Description
    Hardness
    Septum
    Thickness
    Cap colour
    Cap Size
    Material
    Availability
    Price
  • 15
    Magnetic crimp cap
    45 shore
    Blue silicone/clear PTFE, soft
    3,0 mm
    Gold
    20 mm
    Steel
    Log in for availability
    Log in for availability
    1 * 5.000 KS
    1 * 5.000 KS - 181 210,00 Kč
    181 210,00 Kč
    Supplier Article Number: 6AMCC20-ST3F
    Local Article Number: HYPE6AMCC20-ST3F
Product Information
  • Catalog #
    Seq
    Description
    Hardness
    Septum
    Thickness
    Cap colour
    Cap Size
    Material
    Availability
    Price
  • 10
    Magnetic crimp cap
    55 shore
    Red silicone/natural PTFE, high temperature
    3,0 mm
    Silver
    20 mm
    Steel
    Log in for availability
    Log in for availability
    1 * 100 KS
    1 * 100 KS - 3 500,00 Kč
    3 500,00 Kč
    Supplier Article Number: 6AMCC20-STH
    Local Article Number: HYPE6AMCC20-STH
Product Information
  • Catalog #
    Seq
    Description
    Hardness
    Septum
    Thickness
    Cap colour
    Cap Size
    Material
    Availability
    Price
  • 9
    Magnetic crimp cap
    55 shore
    Red silicone/natural PTFE, high temperature
    3,0 mm
    Silver
    20 mm
    Steel
    Log in for availability
    Log in for availability
    1 * 1.000 KS
    1 * 1.000 KS - 33 210,00 Kč
    33 210,00 Kč
    Supplier Article Number: 6AMCC20-STHT
    Local Article Number: HYPE6AMCC20-STHT
Product Information
  • Catalog #
    Seq
    Description
    Hardness
    Septum
    Thickness
    Cap colour
    Cap Size
    Material
    Availability
    Price
  • 12
    Magnetic crimp cap
    55 shore
    Red silicone/natural PTFE, high temperature
    3,0 mm
    Silver
    20 mm
    Steel
    Log in for availability
    Log in for availability
    1 * 5.000 KS
    1 * 5.000 KS - 157 740,00 Kč
    157 740,00 Kč
    Supplier Article Number: 6AMCC20-STHF
    Local Article Number: HYPE6AMCC20-STHF