Tisk…

6-Amino-3-chloropicolinonitrile 95%

BLDPBD00732333-100EA 6790 CZK
BLDPBD00732333-100
6-Amino-3-chloropicolinonitrile 95%
6-Amino-3-chloropicolinonitrile

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Vzorec: C₆H₄ClN₃
MW: 153,5691 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08062817
CAS číslo: 1314976-40-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR