Tisk…

2-Chloro-N,N-dimethylpyrimidin-5-amine 95%

BLDPBD00733084-1GEA 19190 CZK
BLDPBD00733084-1G BLDPBD00733084-250 BLDPBD00733084-100
2-Chloro-N,N-dimethylpyrimidin-5-amine 95%
2-Chloro-N,N-dimethylpyrimidin-5-amine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Vzorec: C₆H₈ClN₃
MW: 157,6008 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD10697652
CAS číslo: 62802-43-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR