Tisk…

3-(Ethylthio)picolinic acid 95%

BLDPBD00737164-1GEA 4960 CZK
BLDPBD00737164-1G BLDPBD00737164-250 BLDPBD00737164-100
3-(Ethylthio)picolinic acid 95%
3-(Ethylthio)picolinic acid

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₈H₉NO₂S
MW: 183,2276 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20647127
CAS číslo: 14440-97-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR