Tisk…

Fmoc-NH-PEG4-CH2CO2H 97%

BLDPBD00737541-250EA 2430 CZK
BLDPBD00737541-250 BLDPBD00737541-5G BLDPBD00737541-1G
Fmoc-NH-PEG4-CH2CO2H 97%
Fmoc-NH-PEG4-CH2CO2H

Storage condition: Sealed in dry, 2-8°C

Vzorec: C₂₅H₃₁NO₈
MW: 473,516 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD26142981
CAS číslo: 437655-95-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR