Tisk…

6-Chloro-1-methylindolin-2-one 95+%

BLDPBD00738629-100EA 1800 CZK
BLDPBD00738629-100 BLDPBD00738629-250 BLDPBD00738629-1G
6-Chloro-1-methylindolin-2-one 95+%
6-Chloro-1-methylindolin-2-one

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₉H₈ClNO
MW: 181,6189 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20483430
CAS číslo: 156136-55-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR