Tisk…

5-Chloro-6-methoxypicolinaldehyde 95%

BLDPBD00743257-250EA 7330 CZK
BLDPBD00743257-250 BLDPBD00743257-1G BLDPBD00743257-100
5-Chloro-6-methoxypicolinaldehyde 95%
5-Chloro-6-methoxypicolinaldehyde

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₇H₆ClNO₂
MW: 171,5810 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16606539
CAS číslo: 1211527-87-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR