Tisk…

N-Carbobenzoxy-L-leucine, ~3% EA 97%

BLDPBD00743494-0.5EA 0 CZK
BLDPBD00743494-0.5 BLDPBD00743494-25G BLDPBD00743494-500 BLDPBD00743494-10G BLDPBD00743494-5G BLDPBD00743494100G BLDPBD007434941000
N-Carbobenzoxy-L-leucine, ~3% EA 97%
N-Carbobenzoxy-L-leucine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Vzorec: C₁₄H₁₉NO₄
MW: 265,31 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00026494
CAS číslo: 2018-66-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR