Tisk…

7-Bromo-4,6-dichloro-2-methylquinazoline 95%

BLDPBD00743728-100EA 1940 CZK
BLDPBD00743728-100 BLDPBD00743728-1G BLDPBD00743728-250
7-Bromo-4,6-dichloro-2-methylquinazoline 95%
7-Bromo-4,6-dichloro-2-methylquinazoline

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₉H₅BrCl₂N₂
MW: 291,9594 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD31698326
CAS číslo: 2384204-25-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR