Tisk…

4,4'-Carbonyldimorpholine 98%

BLDPBD00744091-5GEA 3600 CZK
BLDPBD00744091-5G BLDPBD00744091-250 BLDPBD00744091-25G BLDPBD00744091-1G
4,4'-Carbonyldimorpholine 98%
4,4'-Carbonyldimorpholine

Storage condition: Sealed in dry, 2-8°C

Vzorec: C₉H₁₆N₂O₃
MW: 200,24 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00593814
CAS číslo: 38952-62-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR