Tisk…

2-Methyl-[1,1'-biphenyl]-3-carbaldehyde 95%

BLDPBD00745065-1GEA 2060 CZK
BLDPBD00745065-1G BLDPBD00745065-250
2-Methyl-[1,1'-biphenyl]-3-carbaldehyde 95%
2-Methyl-[1,1'-biphenyl]-3-carbaldehyde

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₁₄H₁₂O
MW: 196,24 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11553605
CAS číslo: 89951-60-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR