Tisk…

3,5-Dichloropyrazolo[1,5-a]pyrimidine 95%

BLDPBD00746709-250EA 2540 CZK
BLDPBD00746709-250 BLDPBD00746709-100 BLDPBD00746709-5G BLDPBD00746709-1G
3,5-Dichloropyrazolo[1,5-a]pyrimidine 95%
3,5-Dichloropyrazolo[1,5-a]pyrimidine

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₆H₃Cl₂N₃
MW: 188,0141 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD26517875
CAS číslo: 1823383-15-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR