Tisk…

5-Fluoro-4-methyl-2-nitropyridine 95%

BLDPBD00747129-1GEA 22830 CZK
BLDPBD00747129-1G BLDPBD00747129-250 BLDPBD00747129-100
5-Fluoro-4-methyl-2-nitropyridine 95%
5-Fluoro-4-methyl-2-nitropyridine
Vzorec: C₆H₅FN₂O₂
MW: 156,11 g/mol
MDL Number: MFCD28798172
CAS číslo: 1628184-28-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR