Tisk…

4-Bromo-1-(4-methoxybenzyl)-3-nitro-1H-pyrazole 95%

BLDPBD00748213-100EA 1800 CZK
BLDPBD00748213-100 BLDPBD00748213-1G BLDPBD00748213-250
4-Bromo-1-(4-methoxybenzyl)-3-nitro-1H-pyrazole 95%
4-Bromo-1-(4-methoxybenzyl)-3-nitro-1H-pyrazole

Storage condition: Sealed in dry, 2-8°C

Vzorec: C₁₁H₁₀BrN₃O₃
MW: 312,1194 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13243213
CAS číslo: 1455366-34-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR