Tisk…

4-Chloroquinoline-7-carbaldehyde 95%

BLDPBD00749310-250EA 7980 CZK
BLDPBD00749310-250 BLDPBD00749310-1G BLDPBD00749310-100
4-Chloroquinoline-7-carbaldehyde 95%
4-Chloroquinoline-7-carbaldehyde

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₁₀H₆ClNO
MW: 191,6137 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD31698485
CAS číslo: 1785002-28-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR