Tisk…

4-Bromoisoquinoline 2-oxide 95%

BLDPBD00750347-5GEA 6050 CZK
BLDPBD00750347-5G BLDPBD00750347-250 BLDPBD00750347-1G
4-Bromoisoquinoline 2-oxide 95%
4-Bromoisoquinoline 2-oxide

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₉H₆BrNO
MW: 224,0500 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00234486
CAS číslo: 3749-21-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR