Tisk…

tert-Butyl ((3S,5R)-5-(hydroxymethyl)pyrrolidin-3-yl)carbamate 95%

BLDPBD00751918-100EA 2630 CZK
BLDPBD00751918-100
tert-Butyl ((3S,5R)-5-(hydroxymethyl)pyrrolidin-3-yl)carbamate 95%
tert-Butyl ((3S,5R)-5-(hydroxymethyl)pyrrolidin-3-yl)carbamate
Vzorec: C₁₀H₂₀N₂O₃
MW: 216,28 g/mol
MDL Number: MFCD08704538
CAS číslo: 1070295-74-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR