Tisk…

[1,1'-Bis(diphenylphosphino)ferrocene]dichlorocobalt(II) 98%

BLDPBD00752487-250EA 485 CZK
BLDPBD00752487-250 BLDPBD00752487-100 BLDPBD00752487-5G BLDPBD00752487-25G BLDPBD00752487-1G
[1,1'-Bis(diphenylphosphino)ferrocene]dichlorocobalt(II) 98%
[1,1'-Bis(diphenylphosphino)ferrocene]dichlorocobalt(II)

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₃₄H₂₀Cl₂CoFeP₂
MW: 676,15 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03427161
CAS číslo: 67292-36-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR