Tisk…

2,6-Diamino-1H-pyrimidin-4-one 97%

BLDPBD00752686-500EA 2110 CZK
BLDPBD00752686-500 BLDPBD00752686-0.5 BLDPBD007526861000 BLDPBD00752686-5G BLDPBD00752686100G BLDPBD00752686-25G
2,6-Diamino-1H-pyrimidin-4-one 97%
2,6-Diamino-1H-pyrimidin-4-one

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Vzorec: C₄H₆N₄O
MW: 126,12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08528919
CAS číslo: 56-06-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR