Tisk…

(4-(Difluoromethoxy)pyridin-2-yl)methanol 95%

BLDPBD00757374-100EA 10320 CZK
BLDPBD00757374-100
(4-(Difluoromethoxy)pyridin-2-yl)methanol 95%
(4-(Difluoromethoxy)pyridin-2-yl)methanol

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₇H₇F₂NO₂
MW: 175,133 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22552054
CAS číslo: 1315361-55-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR