Tisk…

5-Bromo-6-fluoroindoline 97%

BLDPBD00757704-250EA 5050 CZK
BLDPBD00757704-250 BLDPBD00757704-100
5-Bromo-6-fluoroindoline 97%
5-Bromo-6-fluoroindoline

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Vzorec: C₈H₇BrFN
MW: 216,0503 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22037765
CAS číslo: 1368323-85-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR