Tisk…

3-Bromo-6-cyclopropylpyridazine Hydrobromide 95%

BLDPBD00760907-1GEA 13970 CZK
BLDPBD00760907-1G BLDPBD00760907-100 BLDPBD00760907-250
3-Bromo-6-cyclopropylpyridazine Hydrobromide 95%
3-Bromo-6-cyclopropylpyridazine Hydrobromide

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₇H₈Br₂N₂
MW: 279,9598 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD30344702
CAS číslo: 2044706-90-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR