Tisk…

2,5-Dimethoxypyridine 95%

BLDPBD00761618-1GEA 599 CZK
BLDPBD00761618-1G BLDPBD00761618-25G BLDPBD00761618-5G BLDPBD00761618-250
2,5-Dimethoxypyridine 95%
2,5-Dimethoxypyridine

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₇H₉NO₂
MW: 139,152 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12546620
CAS číslo: 867267-24-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR