Tisk…

4-Fluorothieno[2,3-c]pyridine-2-carboxylic acid 95%

BLDPBD00761949-1GEA 11800 CZK
BLDPBD00761949-1G BLDPBD00761949-100 BLDPBD00761949-250
4-Fluorothieno[2,3-c]pyridine-2-carboxylic acid 95%
4-Fluorothieno[2,3-c]pyridine-2-carboxylic acid

Storage condition: Sealed in dry, 2-8°C

Vzorec: C₈H₄FNO₂S
MW: 197,1800 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD20657771
CAS číslo: 870235-46-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR