Tisk…

Methyl 6-fluoro-5-(tetrahydrofuran-3-yl)nicotinate 97%

BLDPBD00761969-100EA 2740 CZK
BLDPBD00761969-100 BLDPBD00761969-50M
Methyl 6-fluoro-5-(tetrahydrofuran-3-yl)nicotinate 97%
Methyl 6-fluoro-5-(tetrahydrofuran-3-yl)nicotinate

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₁₁H₁₂FNO₃
MW: 225,2163 g/mol
Storage Temperature: Ambient
CAS číslo: 2288708-92-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR