Tisk…

3-Bromo-5-chloropyridin-4(1H)-one 95%

BLDPBD00763126-250EA 2570 CZK
BLDPBD00763126-250 BLDPBD00763126-100 BLDPBD00763126-1G
3-Bromo-5-chloropyridin-4(1H)-one 95%
3-Bromo-5-chloropyridin-4(1H)-one

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₅H₃BrClNO
MW: 208,4404 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13185351
CAS číslo: 70149-40-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR