Tisk…

3-Fluoro-2-methoxyisonicotinic acid 95%

BLDPBD00763356-1GEA 9780 CZK
BLDPBD00763356-1G BLDPBD00763356-100 BLDPBD00763356-5G BLDPBD00763356-250
3-Fluoro-2-methoxyisonicotinic acid 95%
3-Fluoro-2-methoxyisonicotinic acid

Storage condition: Sealed in dry, 2-8°C

Vzorec: C₇H₆FNO₃
MW: 171,1300 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13185871
CAS číslo: 1214362-24-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR