Tisk…

(Triphenylphosphonio)difluoroacetate 95%

BLDPBD00763988-5GEA 6250 CZK
BLDPBD00763988-5G BLDPBD00763988-1G BLDPBD00763988-100 BLDPBD00763988-250
(Triphenylphosphonio)difluoroacetate 95%
(Triphenylphosphonio)difluoroacetate

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₂₀H₁₅F₂O₂P
MW: 356,302 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28359690
CAS číslo: 1449521-05-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR