Tisk…

2-Ethynyl-3-fluoropyridine 98%

BLDPBD00764736-100EA 941 CZK
BLDPBD00764736-100 BLDPBD00764736-250 BLDPBD00764736-1G
2-Ethynyl-3-fluoropyridine 98%
2-Ethynyl-3-fluoropyridine

Storage condition: Sealed in dry, 2-8°C

Vzorec: C₇H₄FN
MW: 121,1118 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22191371
CAS číslo: 1401685-43-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR