Tisk…

[(2-chloro-4-fluorophenyl)methyl]hydrazine dihydrochloride 97%

BLDPBD00764766-5GEA 7410 CZK
BLDPBD00764766-5G BLDPBD00764766-1G BLDPBD00764766-250
[(2-chloro-4-fluorophenyl)methyl]hydrazine dihydrochloride 97%
[(2-chloro-4-fluorophenyl)methyl]hydrazine dihydrochloride

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₇H₁₀Cl₃FN₂
MW: 247,5251 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD30489782
CAS číslo: 2044707-02-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR