Tisk…

(1S,2S)-2-Aminocyclohexane-1-carboxylic acid hydrochloride 95%

BLDPBD00765028-1GEA 19070 CZK
BLDPBD00765028-1G BLDPBD00765028-250 BLDPBD00765028-100
(1S,2S)-2-Aminocyclohexane-1-carboxylic acid hydrochloride 95%
(1S,2S)-2-Aminocyclohexane-1-carboxylic acid hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₇H₁₄ClNO₂
MW: 179,6446 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13195001
CAS číslo: 24716-92-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR