Tisk…

Ethyl 8-bromo-1,6-naphthyridine-3-carboxylate 97%

BLDPBD00765175-100EA 10040 CZK
BLDPBD00765175-100 BLDPBD00765175-250 BLDPBD00765175-1G
Ethyl 8-bromo-1,6-naphthyridine-3-carboxylate 97%
Ethyl 8-bromo-1,6-naphthyridine-3-carboxylate

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₁₁H₉BrN₂O₂
MW: 281,1054 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30828915
CAS číslo: 2089652-13-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR