Tisk…

Methyl 2-thioxo-2,3-dihydro-1H-benzo[d]imidazole-5-carboxylate 98%

BLDPBD00765659-100EA 2400 CZK
BLDPBD00765659-100 BLDPBD00765659-250
Methyl 2-thioxo-2,3-dihydro-1H-benzo[d]imidazole-5-carboxylate 98%
Methyl 2-thioxo-2,3-dihydro-1H-benzo[d]imidazole-5-carboxylate

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₉H₈N₂O₂S
MW: 208,2370 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09192253
CAS číslo: 64375-41-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR