Tisk…

Methyl 6-bromo-2-methyl-2H-indazole-3-carboxylate 98+%

BLDPBD00767129-250EA 11800 CZK
BLDPBD00767129-250 BLDPBD00767129-100 BLDPBD00767129-1G
Methyl 6-bromo-2-methyl-2H-indazole-3-carboxylate 98+%
Methyl 6-bromo-2-methyl-2H-indazole-3-carboxylate

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₁₀H₉BrN₂O₂
MW: 269,0947 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14584976
CAS číslo: 1216852-55-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR