Tisk…

Methyl 5-(bromomethyl)-2-nitrobenzoate 98%

BLDPBD00767596-1GEA 1290 CZK
BLDPBD00767596-1G BLDPBD00767596-250 BLDPBD00767596-5G
Methyl 5-(bromomethyl)-2-nitrobenzoate 98%
Methyl 5-(bromomethyl)-2-nitrobenzoate

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₉H₈BrNO₄
MW: 274,0681 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD20528191
CAS číslo: 88071-91-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR