Tisk…

(2-Amino-5-(3-(piperidin-1-yl)propoxy)phenyl)methanol 97%

BLDPBD00768620-50MEA 1430 CZK
BLDPBD00768620-50M BLDPBD00768620-5G BLDPBD00768620-250 BLDPBD00768620-1G BLDPBD00768620-100
(2-Amino-5-(3-(piperidin-1-yl)propoxy)phenyl)methanol 97%
(2-Amino-5-(3-(piperidin-1-yl)propoxy)phenyl)methanol

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Vzorec: C₁₅H₂₄N₂O₂
MW: 264,3633 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31715177
CAS číslo: 2288710-47-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR