Tisk…

Fmoc-HomoSer(TBDMS)-OH 95%

BLDPBD00770875-1GEA 2480 CZK
BLDPBD00770875-1G BLDPBD00770875-250
Fmoc-HomoSer(TBDMS)-OH 95%
Fmoc-HomoSer(TBDMS)-OH
Vzorec: C₂₅H₃₃NO₅Si
MW: 455,6 g/mol
CAS číslo: 1333332-17-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR