Tisk…

5-Chloro-2,4-difluoropyridine 95%

BLDPBD00772131-1GEA 34120 CZK
BLDPBD00772131-1G BLDPBD00772131-100 BLDPBD00772131-50M BLDPBD00772131-250
5-Chloro-2,4-difluoropyridine 95%
5-Chloro-2,4-difluoropyridine

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₅H₂ClF₂N
MW: 149,5259 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22577173
CAS číslo: 1807257-52-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR