Tisk…

Methyl 6-bromopyrazolo[1,5-a]pyrimidine-3-carboxylate 95%

BLDPBD00775255-100EA 1320 CZK
BLDPBD00775255-100 BLDPBD00775255-250 BLDPBD00775255-1G
Methyl 6-bromopyrazolo[1,5-a]pyrimidine-3-carboxylate 95%
Methyl 6-bromopyrazolo[1,5-a]pyrimidine-3-carboxylate

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₈H₆BrN₃O₂
MW: 256,0561 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13194149
CAS číslo: 1263060-83-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR