Tisk…

3-(3-Bromopropyl)thiophene 95%

BLDPBD00775696-1GEA 25830 CZK
BLDPBD00775696-1G BLDPBD00775696-250 BLDPBD00775696-100
3-(3-Bromopropyl)thiophene 95%
3-(3-Bromopropyl)thiophene

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Vzorec: C₇H₉BrS
MW: 205,1154 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16824798
CAS číslo: 121459-86-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR