Tisk…

4-Methoxy-5-(3-morpholinopropoxy)-2-nitrobenzamide 95%

BLDPBD00777310-250EA 200 CZK
BLDPBD00777310-250 BLDPBD00777310-5G BLDPBD00777310-1G
4-Methoxy-5-(3-morpholinopropoxy)-2-nitrobenzamide 95%
4-Methoxy-5-(3-morpholinopropoxy)-2-nitrobenzamide

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Vzorec: C₁₅H₂₁N₃O₆
MW: 339,3437 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD31579912
CAS číslo: 861453-16-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR