Tisk…

Ethyl 1-methyl-1H-pyrrolo[3,2-b]pyridine-6-carboxylate 95%

BLDPBD00778800-1GEA 9980 CZK
BLDPBD00778800-1G BLDPBD00778800-250 BLDPBD00778800-5G BLDPBD00778800-100
Ethyl 1-methyl-1H-pyrrolo[3,2-b]pyridine-6-carboxylate 95%
Ethyl 1-methyl-1H-pyrrolo[3,2-b]pyridine-6-carboxylate

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₁₁H₁₂N₂O₂
MW: 204,2252 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31705012
CAS číslo: 2114651-27-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR