Tisk…

2-Chloro-4-iodo-3-methylpyridine 97%

BLDPBD00779759-5GEA 3080 CZK
BLDPBD00779759-5G BLDPBD00779759-25G BLDPBD00779759-250 BLDPBD00779759-1G
2-Chloro-4-iodo-3-methylpyridine 97%
2-Chloro-4-iodo-3-methylpyridine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Vzorec: C₆H₅ClIN
MW: 253,47 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03095293
CAS číslo: 153034-88-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR