Tisk…

(4-(Bis(benzo[d][1,3]dioxol-5-yl)methyl)piperidin-1-yl)(1H-1,2,4-triazol-1-yl)methanone 98%

BLDPBD00782109-5MGEA 4020 CZK
BLDPBD00782109-5MG BLDPBD00782109-10M BLDPBD00782109-25M
(4-(Bis(benzo[d][1,3]dioxol-5-yl)methyl)piperidin-1-yl)(1H-1,2,4-triazol-1-yl)methanone 98%
(4-(Bis(benzo[d][1,3]dioxol-5-yl)methyl)piperidin-1-yl)(1H-1,2,4-triazol-1-yl)methanone

Storage condition: Sealed in dry, Store in freezer, under -20°C

Vzorec: C₂₃H₂₂N₄O₅
MW: 434,4446 g/mol
Storage Temperature: Freezer
MDL Number: MFCD30182268
CAS číslo: 1515855-97-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR