Tisk…

Gallium(III) acetylacetonate 99.99% (metals basis)

BLDPBD00782821-25GEA 5390 CZK
BLDPBD00782821-25G BLDPBD00782821-5G BLDPBD00782821-1G
Gallium(III) acetylacetonate 99.99% (metals basis)
Gallium(III) acetylacetonate

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₁₅H₂₁GaO₆
MW: 367,05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00013492
CAS číslo: 14405-43-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR