Tisk…

N-(5-Chloropyridin-2-yl)-2-(4-(N,N-dimethylcarbamimidoyl)benzamido)-5-methoxybenzamide maleate 98+%

BLDPBD00785609-100EA 2030 CZK
BLDPBD00785609-100 BLDPBD00785609-1G BLDPBD00785609-250
N-(5-Chloropyridin-2-yl)-2-(4-(N,N-dimethylcarbamimidoyl)benzamido)-5-methoxybenzamide maleate 98+%
N-(5-Chloropyridin-2-yl)-2-(4-(N,N-dimethylcarbamimidoyl)benzamido)-5-methoxybenzamide maleate

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Vzorec: C₂₇H₂₆ClN₅O₇
MW: 567,9777 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31729381
CAS číslo: 936539-80-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR