Tisk…

1-Ethyl-7-methoxy-1,2,3,4-tetrahydroquinoline 97%

BLDPBD00785660-100EA 998 CZK
BLDPBD00785660-100
1-Ethyl-7-methoxy-1,2,3,4-tetrahydroquinoline 97%
1-Ethyl-7-methoxy-1,2,3,4-tetrahydroquinoline

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₁₂H₁₇NO
MW: 191,2695 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31715241
CAS číslo: 150749-32-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR