Tisk…

2,6-Dibromo-4-methylanisole 98%

BLDPBD00785864-1GEA 1970 CZK
BLDPBD00785864-1G BLDPBD00785864-0.0 BLDPBD00785864-25G BLDPBD00785864-5G
2,6-Dibromo-4-methylanisole 98%
2,6-Dibromo-4-methylanisole

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₈H₈Br₂O
MW: 279,96 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00196821
CAS číslo: 51699-89-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR