Tisk…

2H-1,2,3,4-tetrazole-5-carboxylic acid 95%

BLDPBD00786509-250EA 2970 CZK
BLDPBD00786509-250 BLDPBD00786509-100 BLDPBD00786509-1G BLDPBD00786509-5G
2H-1,2,3,4-tetrazole-5-carboxylic acid 95%
2H-1,2,3,4-tetrazole-5-carboxylic acid

Storage condition: Sealed in dry, 2-8°C

Vzorec: C₂H₂N₄O₂
MW: 114,0629 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD17676507
CAS číslo: 75773-99-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR