Tisk…

7-Bromo-2-chloroquinazolin-4-ol 95%

BLDPBD00786672-250EA 4510 CZK
BLDPBD00786672-250 BLDPBD00786672-1G BLDPBD00786672-100
7-Bromo-2-chloroquinazolin-4-ol 95%
7-Bromo-2-chloroquinazolin-4-ol

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₈H₄BrClN₂O
MW: 259,4872 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD30299670
CAS číslo: 2349411-92-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR